Phenazine is the parent substance of many dyestuffs, such as the toluylene red, indulines and safranines. It can be applied in therapeutic uses, used as antipsychotic and antiemetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
Morimitsu Nishikimi.; N. Appaji Rao.; Kunio Yagi. The occurrence of superoxide anion in the reaction of reduced phenazine methosulfate and molecular oxygen. Biochem. Biophys. Res. Commun. 1972, 46 (2), 849-854 .
Korneel Rabaey.; Nico Boon.; Monica Höfte .; Willy Verstraete. Microbial Phenazine Production Enhances Electron Transfer in Biofuel Cells.Environ. Sci. Technol. 2005, 39 (9),3401-3408 .