Thermo Scientific Chemicals

Phenazine, 99+%, Thermo Scientific Chemicals

Catalog number: A15770.14
25 g, Each
Thermo Scientific Chemicals

Phenazine, 99+%, Thermo Scientific Chemicals

Catalog number: A15770.14
25 g, Each
Quantity
Catalog number: A15770.14
also known as A15770-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92-82-0
IUPAC Name
phenazine
Molecular Formula
C12H8N2
InChI Key
PCNDJXKNXGMECE-UHFFFAOYSA-N
SMILES
C1=CC2=NC3=CC=CC=C3N=C2C=C1
Appearance (Color)
Yellow to yellow-green
Form
Crystals or powder or crystalline powder or needles
Identification (FTIR)
Conforms
Melting Point (clear melt)
171.0-177.0?C
Assay (GC)
≥99.0%

Description

Phenazine is the parent substance of many dyestuffs, such as the toluylene red, indulines and safranines. It can be applied in therapeutic uses, used as antipsychotic and antiemetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text