Thermo Scientific Chemicals

2,2'-Bipyridine, 99+%, Thermo Scientific Chemicals

Catalog number: A15782.36
500 g, Each
Thermo Scientific Chemicals

2,2'-Bipyridine, 99+%, Thermo Scientific Chemicals

Catalog number: A15782.36
500 g, Each
Quantity
Catalog number: A15782.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
366-18-7
IUPAC Name
2,2'-bipyridine
Molecular Formula
C10H8N2
InChI Key
ROFVEXUMMXZLPA-UHFFFAOYSA-N
SMILES
C1=CC=C(N=C1)C1=CC=CC=N1
Identification (FTIR)
Conforms
Melting Point (clear melt)
67-73?C
Appearance (Color)
White to pale cream or pale pink
Form
Crystalline powder or crystals
Assay (GC)
≥99.0%

Description

Reagent for iron determinations.2,2'-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for iron determinations.2,2′-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.

Solubility
Soluble in organic solvents and slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text