Diphenyl ether, 99%, Thermo Scientific Chemicals
Diphenyl ether, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenyl ether, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
250 g
500 g
Catalog number A15791.0E
also known as A15791-0E
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
Add to cart
Diphenyl ether, 99%, Thermo Scientific Chemicals
Catalog numberA15791.0E
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS101-84-8
IUPAC Namephenoxybenzene
Molecular FormulaC12H10O
InChI KeyUSIUVYZYUHIAEV-UHFFFAOYSA-N
SMILESO(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5790-1.5820 @ 20?C
Diphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.

Solubility
Soluble in alcohol, chloroform, ethyl acetate, benzene, diethyl ether and glacial acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Useful reaction medium for high-temperature reactions, e.g. cyclizations and decarboxylations.
  2. Yokota, N.; Ono, H.; Miyake, J.; Nishino, E.; Asazawa, K.; Watanabe, M.; Miyatake, K. Anion Conductive Aromatic Block Copolymers Containing Diphenyl Ether or Sulfide Groups for Application to Alkaline Fuel Cells. ACS Appl. Mater. Interfaces 2014, 6 (19), 17044-17052.
  3. Stelt, T. P.; Casati, E.; Colonna, N. C. P. Technical equation of state models for heat transfer fluids made of biphenyl and diphenyl ether and their mixtures. Fluid Phase Equilib. 2015, 393, 64-77.