Thermo Scientific Chemicals

Diphenyl ether, 99%, Thermo Scientific Chemicals

Catalog number: A15791.36
500 g, Each
Thermo Scientific Chemicals

Diphenyl ether, 99%, Thermo Scientific Chemicals

Catalog number: A15791.36
500 g, Each
Quantity
Catalog number: A15791.36
also known as A15791-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101-84-8
IUPAC Name
phenoxybenzene
Molecular Formula
C12H10O
InChI Key
USIUVYZYUHIAEV-UHFFFAOYSA-N
SMILES
O(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5790-1.5820 @ 20?C

Description

Diphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.

Solubility
Soluble in alcohol, chloroform, ethyl acetate, benzene, diethyl ether and glacial acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text