Thermo Scientific Chemicals

6-Hydroxynicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15802.14
25 g, Each
Thermo Scientific Chemicals

6-Hydroxynicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15802.14
25 g, Each
Quantity
Catalog number: A15802.14
also known as A15802-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5006-66-6
IUPAC Name
6-oxo-1,6-dihydropyridine-3-carboxylic acid
Molecular Formula
C6H5NO3
InChI Key
BLHCMGRVFXRYRN-UHFFFAOYSA-N
SMILES
OC(=O)C1=CNC(=O)C=C1
Form
Powder
Appearance (Color)
Pale cream to cream to brown
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (HPLC)
≥97.5%

Description

Metabolite of nicotinamide. Stimulates adrenocortical secretion

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text