Thermo Scientific Chemicals

4-Hydroxybenzonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A15803.22
100 g, Each
Thermo Scientific Chemicals

4-Hydroxybenzonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A15803.22
100 g, Each
Quantity
Catalog number: A15803.22
also known as A15803-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
767-00-0
IUPAC Name
4-hydroxybenzonitrile
Molecular Formula
C7H5NO
InChI Key
CVNOWLNNPYYEOH-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C#N
Appearance (Color)
White to cream
Form
Powder
Melting Point (clear melt)
110-115?C
Assay (GC)
≥98.0%

Description

4-Hydroxybenzonitrile is used as an intermediate in organic synthesis. It is also used in cosmetic as an antiseptic and disinfectant. It is involved as a raw material for the preparation of bomoxynil and ioxynil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzonitrile is used as an intermediate in organic synthesis. It is also used in cosmetic as an antiseptic and disinfectant. It is involved as a raw material for the preparation of bomoxynil and ioxynil.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text