4-Hydroxybenzonitrile, 98+%, Thermo Scientific Chemicals
4-Hydroxybenzonitrile, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxybenzonitrile, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
100 g
250 g
Catalog number A15803.36
also known as A15803-36
Price (USD)/ Each
490.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
490.00
Add to cart
4-Hydroxybenzonitrile, 98+%, Thermo Scientific Chemicals
Catalog numberA15803.36
Price (USD)/ Each
490.00
-
Add to cart
Chemical Identifiers
CAS767-00-0
IUPAC Name4-hydroxybenzonitrile
Molecular FormulaC7H5NO
InChI KeyCVNOWLNNPYYEOH-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Melting Point (clear melt)110-115?C
Assay (GC)≥98.0%
4-Hydroxybenzonitrile is used as an intermediate in organic synthesis. It is also used in cosmetic as an antiseptic and disinfectant. It is involved as a raw material for the preparation of bomoxynil and ioxynil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzonitrile is used as an intermediate in organic synthesis. It is also used in cosmetic as an antiseptic and disinfectant. It is involved as a raw material for the preparation of bomoxynil and ioxynil.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Durgannavar, A. K.; Patgar, M. B.; Chimatadar, S. A. Oxidation of amoxicillin by hexacyanoferrate(III) in aqueous alkaline medium-A kinetic and mechanistic approach. Indian J Chem 2015, 54 (9), 1085-1091.
  2. Zhang, J.; Wu, W.; Ji, X.; Cao, S. Perfluorobutyl iodide-assisted direct cyanomethylation of azoles and phenols with acetonitrile. RSC Adv. 2015, 5 (26), 20562-20565.