L-Alanine, 99%
Thermo Scientific Chemicals

L-Alanine, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A15804.36
also known as A15804-36
Price (USD)/ Each
286.65
Online exclusive
318.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
286.65
Online exclusive
318.00 
Save 31.35 (10%)
Add to cart
L-Alanine, 99%
Catalog numberA15804.36
Price (USD)/ Each
286.65
Online exclusive
318.00 
Save 31.35 (10%)
-
Add to cart
Chemical Identifiers
CAS56-41-7
IUPAC Name(2S)-2-aminopropanoic acid
Molecular FormulaC3H7NO2
InChI KeyQNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILESC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
CommentSpecification differs for U.S. and non-U.S. material where indicated
Optical Rotation+13.7 to +15.1 (U.S. specification)
Appearance (Color)White
Assay from Suppliers CofA≥98.5% (U.S. specification)
View more
L-Alanine plays a vital role in glucose-alanine cycle between tissue and liver. It is used for protein construction as well as involved in the metabolism of tryptophan and vitamin pyridoxine. It provides energy for muscles and central nervous system in order to strengthen the immune system. It helps in the metabolism of sugars and organic acids, and enhances the germination rates of Bacillus subtilis spores. It also displays a cholesterol-reducing effect in animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and cold ethanol. Slightly soluble in pyridine. Insoluble in ether and acetone.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. ɑ-Amino acids can be converted to the corresponding ɑ-chloro acids with a high degree of retention of configuration, by diazotization in the presence of HCl: J. Am. Chem. Soc., 76, 6054 (1954). The intermediate is thought to be a rigid oxiranone which prevents racemization. For an improved technique and list of examples, see: Org. Synth. Coll., 8, 119 (1993):
  2. Moozeh, K.; So, M. S.; Chin, J. Catalytic Stereoinversion of L-Alanine to Deuterated D-Alanine. Angew. Chem. 2015, 127 (32), 9513-9517.
  3. Richter, N.; Farnberger, J. E.; Pressnitz, D.; Lechner, H.; Zepeck, F.; Kroutil, W. A system for μ-transaminase mediated (R)-amination using l-alanine as an amine donor. Green Chem. 2015, 17 (5), 2952-2958.