Cadmium carbonate, 97%, Thermo Scientific Chemicals
Cadmium carbonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium carbonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A15812.0B
also known as A15812-0B
Price (USD)/ Each
-
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS513-78-0
IUPAC Namecadmium(2+) carbonate
Molecular FormulaCCdO3
InChI KeyGKDXQAKPHKQZSC-UHFFFAOYSA-L
SMILES[Cd++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Complexometric Titration≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only