Thermo Scientific Chemicals

(±)-Epichlorohydrin, 99%, Thermo Scientific Chemicals

Catalog number: A15823.22
100 g, Each
Thermo Scientific Chemicals

(±)-Epichlorohydrin, 99%, Thermo Scientific Chemicals

Catalog number: A15823.22
100 g, Each
Quantity
Catalog number: A15823.22
also known as A15823-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-89-8
IUPAC Name
2-(chloromethyl)oxirane
Molecular Formula
C3H5ClO
InChI Key
BRLQWZUYTZBJKN-UHFFFAOYNA-N
SMILES
ClCC1CO1
Refractive Index
1.4365-1.4395 @ 20°C (UK-sourced material)
Comment
Material Sourced in UK and US
Appearance (Color)
Clear colorless
Form
Liquid
Assay from Supplier's CofA
≥98.5% (US-sourced material)

Description

Epichlorohydrin is used in the manufacture of polyamines and polyquaternary ammonium salts, as flocculents in water and wastewater treatment. It is used in the synthesis of surface active agents for washing products and toiletries. Polyamine-epichlorohydrin resin is used in paper industry to improve paper wet-strength. It acts as a chemical intermediate and widely used in the manufacture of epoxy resins, epichlorohydrin elastomers and polyamide-epichlorohydrin resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Epichlorohydrin is used in the manufacture of polyamines and polyquaternary ammonium salts, as flocculents in water and wastewater treatment. It is used in the synthesis of surface active agents for washing products and toiletries. Polyamine-epichlorohydrin resin is used in paper industry to improve paper wet-strength. It acts as a chemical intermediate and widely used in the manufacture of epoxy resins, epichlorohydrin elastomers and polyamide-epichlorohydrin resins.

Solubility
Miscible with water and organic polar solvents.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text