2-Chlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals
2-Chlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A15826.22
also known as A15826-22
Price (USD)/ Each
241.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
241.00
Add to cart
2-Chlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals
Catalog numberA15826.22
Price (USD)/ Each
241.00
-
Add to cart
Chemical Identifiers
CAS39718-00-8
IUPAC Name1-chloro-2-(methylsulfanyl)benzene
Molecular FormulaC7H7ClS
InChI KeyIHLDFHCSSCVPQW-UHFFFAOYSA-N
SMILESCSC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5885-1.5935 @ 20?C
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only