Thermo Scientific Chemicals

tert-Butyl chloroacetate, 98%, Thermo Scientific Chemicals

Catalog number: A15833.36
500 g, Each
Thermo Scientific Chemicals

tert-Butyl chloroacetate, 98%, Thermo Scientific Chemicals

Catalog number: A15833.36
500 g, Each
Quantity
Catalog number: A15833.36
also known as A15833-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-59-5
IUPAC Name
tert-butyl 2-chloroacetate
Molecular Formula
C6H11ClO2
InChI Key
KUYMVWXKHQSIAS-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)CCl
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4215-1.4255 @ 20°C
Assay (GC)
≥96.0%

Description

tert-Butyl chloroacetate is used in the synthesis of imidazol-1-yl-acetic acid hydrochloride, cis-disubstituted aziridine ester via aza-Darzens reaction and 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl chloroacetate is used in the synthesis of imidazol-1-yl-acetic acid hydrochloride, cis-disubstituted aziridine ester via aza-Darzens reaction and 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore.

Solubility
Decomposes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text