2-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals
2-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number A15834.30
also known as A15834-30
Price (USD)/ Each
446.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
446.00
Add to cart
2-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals
Catalog numberA15834.30
Price (USD)/ Each
446.00
-
Add to cart
Chemical Identifiers
CAS2444-36-2
IUPAC Name2-(2-chlorophenyl)acetic acid
Molecular FormulaC8H7ClO2
InChI KeyIUJAAIZKRJJZGQ-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Appearance (Color)White to pale yellow
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)93.0-99.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only