Thermo Scientific Chemicals

3-Chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: A15844.18
50 g, Each
Thermo Scientific Chemicals

3-Chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: A15844.18
50 g, Each
Quantity
Catalog number: A15844.18
also known as A15844-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
766-80-3
IUPAC Name
1-(bromomethyl)-3-chlorobenzene
Molecular Formula
C7H6BrCl
InChI Key
LZIYAIRGDHSVED-UHFFFAOYSA-N
SMILES
ClC1=CC=CC(CBr)=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5860-1.5910 @ 20?C

Description

3-Chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers. It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole dihydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers. It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole dihydrochloride.

Solubility
Decomposes in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text