Methyl formate, 97%, may cont. up to ca 3% methanol, Thermo Scientific Chemicals
Methyl formate, 97%, may cont. up to ca 3% methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl formate, 97%, may cont. up to ca 3% methanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
500 mL
10,000 mL
Catalog number A15850.0F
also known as A15850-0F
Price (USD)/ Each
62.65
Online exclusive
68.90 
Save 6.25 (9%)
-
Add to cart
Quantity:
2500 mL
Request bulk or custom format
Price (USD)/ Each
62.65
Online exclusive
68.90 
Save 6.25 (9%)
Add to cart
Methyl formate, 97%, may cont. up to ca 3% methanol, Thermo Scientific Chemicals
Catalog numberA15850.0F
Price (USD)/ Each
62.65
Online exclusive
68.90 
Save 6.25 (9%)
-
Add to cart
Chemical Identifiers
CAS107-31-3
IUPAC Namemethyl formate
Molecular FormulaC2H4O2
InChI KeyTZIHFWKZFHZASV-UHFFFAOYSA-N
SMILESCOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.3415-1.3465 @ 20?C
Assay (GC)≥96.0%
Methyl formate is used as a precursor to formamide, dimethylformamide and formic acid. It is used as a blowing agent and utilized for quick-drying finishes due to its high vapor pressure. It is used as a component of azeotrope to recover expensive solvents. Further, it is used as a propellant and an intermediate for optical brighteners and hydrocyanic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl formate is used as a precursor to formamide, dimethylformamide and formic acid. It is used as a blowing agent and utilized for quick-drying finishes due to its high vapor pressure. It is used as a component of azeotrope to recover expensive solvents. Further, it is used as a propellant and an intermediate for optical brighteners and hydrocyanic acid.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Yalfani, M. S.; Lolli, G.; Müller, T. E.; Wolf, A.; Mleczko, L. Methyl N-Phenyl Carbamate Synthesis from Aniline and Methyl Formate: Carbon Recycling to Chemical Products. ChemSusChem 2015, 8 (3), 443-447.
  2. Wu, C.; Zhang, Z.; Zhu, Q.; Han, H.; Yang, Y.; Han, B. Highly efficient hydrogenation of carbon dioxide to methyl formate over supported gold catalysts. Green Chem. 2015, 17 (3), 1467-1472.