Thermo Scientific Chemicals

Cyclohexylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A15851.AP
500 mL, Each
Thermo Scientific Chemicals

Cyclohexylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A15851.AP
500 mL, Each
Quantity
Catalog number: A15851.AP
also known as A15851-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-91-8
IUPAC Name
cyclohexanamine
Molecular Formula
C6H13N
InChI Key
PAFZNILMFXTMIY-UHFFFAOYSA-N
SMILES
NC1CCCCC1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms (non-U.S. specification)

Description

Cyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.

Solubility
Miscible with water, ethanol, ethers, acetone, alcohol and ketones.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, carbon dioxide, sodium hypochlorite, organic acids, mineral acids and peroxides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text