Thermo Scientific Chemicals

4-Hydroxyquinoline, 98%, Thermo Scientific Chemicals

Catalog number: A15859.09
10 g, Each
Thermo Scientific Chemicals

4-Hydroxyquinoline, 98%, Thermo Scientific Chemicals

Catalog number: A15859.09
10 g, Each
Quantity
Catalog number: A15859.09
also known as A15859-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-36-9
Appearance (Color)
White to pale yellow or cream to pale brown
Melting Point (clear melt)
202-206?C
Assay from Supplier's CofA
≥97.5%
Form
Crystals or needles or powder or crystalline powder

Description

4-Hydroxyquinoline is a medical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxyquinoline is a medical raw material.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text