Thermo Scientific Chemicals

Tetraphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A15860.22
100 g, Each
Thermo Scientific Chemicals

Tetraphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A15860.22
100 g, Each
Quantity
Catalog number: A15860.22
also known as A15860-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2751-90-8
IUPAC Name
tetraphenylphosphanium bromide
Molecular Formula
C24H20BrP
InChI Key
BRKFQVAOMSWFDU-UHFFFAOYSA-M
SMILES
[Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream
Water Content (Karl Fischer Titration)
≤1.5%
Assay (Titration ex Bromide)
≥98.0 to ≤102.0%

Description

Tetraphenylphosphonium bromide is a supporting electrolyte for the electroreduction of buckminsterfullerene. It is used to extract heavy metals from aqueous solution as ion-association complexes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylphosphonium bromide is a supporting electrolyte for the electroreduction of buckminsterfullerene. It is used to extract heavy metals from aqueous solution as ion-association complexes

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Incompatible with strong oxidizing agents. Stable at room temperature in closed containers under normal storage and handling conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text