Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A15864.06
5 g, Each
Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A15864.06
5 g, Each
Quantity
Catalog number: A15864.06
also known as A15864-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1620-98-0
IUPAC Name
3,5-di-tert-butyl-4-hydroxybenzaldehyde
Molecular Formula
C15H22O2
InChI Key
DOZRDZLFLOODMB-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC(C=O)=CC(=C1O)C(C)(C)C
Appearance (Color)
Pale cream or pale yellow to yellow
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder or flakes
Melting Point (clear melt)
185-193?C

Description

3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.

Solubility
Soluble in hot methanol.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text