Thermo Scientific Chemicals

Methyltriphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A15878.18
50 g, Each
Thermo Scientific Chemicals

Methyltriphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A15878.18
50 g, Each
Quantity
Catalog number: A15878.18
also known as A15878-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1779-49-3
IUPAC Name
methyltriphenylphosphanium bromide
Molecular Formula
C19H18BrP
InChI Key
LSEFCHWGJNHZNT-UHFFFAOYSA-M
SMILES
[Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Assay (Titration ex Bromide)
≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)
≤1.5%
Form
Crystals or powder or crystalline powder or granules
Appearance (Color)
White
Melting Point (clear melt)
229-238?C

Description

Methyltriphenylphosphonium bromide is used for methylenation through the Wittig reaction. It is utilized in the synthesis of an enyne and 9-isopropenyl -phenanthrene by using sodium amide as reagent. It is a lipophilic molecule and used to deliver molecules to specific cell components. Further, it acts as an antineoplastic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriphenylphosphonium bromide is used for methylenation through the Wittig reaction. It is utilized in the synthesis of an enyne and 9-isopropenyl -phenanthrene by using sodium amide as reagent. It is a lipophilic molecule and used to deliver molecules to specific cell components. Further, it acts as an antineoplastic agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text