The PolarScreen™ ER (Estrogen Receptor) Alpha Competitor Assay Kit, Green, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential ER alpha ligands. The kit uses insect cell-expressed, full-length, native (untagged) human estrogen receptor alpha (available separately, Cat. No. A15674) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone ES2 Green) in a homogenous, mix-and-read assay format.
This kit produces a green color read-out; a 'red' kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance contact drugdiscoverytech@lifetech.com.
PolarScreen™ ER Alpha Competitor Assay, Green, is:
• Complete & ready-to-use out of the box—all reagents included, just add your test compounds • High throughput compatible—800 x 20 μL fluorescence polarization assays in a 384-well plate • Convenient—available in bulk sizes for larger screening needs
How the Assay Works Full length ER alpha is added to a fluorescent estrogen ligand to form an ER-Fluormone ES2 complex. This complex is added to individual test compounds in multiwell plates. To determine relative affinity, the assay measures fluorescence polarization. Competing test compounds will displace the Fluormone ES2 ligand from ER alpha, permitting it to tumble rapidly and resulting in low polarization values. A shift in the fluorescence polarization value in the presence of test compound is used to determine the relative affinity of test compounds for ER alpha.
All components are manufactured under the strictest quality parameters. This product's performance, per the Certificate of Analysis, is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Method
Fluorescence
For Use With (Equipment)
Fluorescent Imager
Format
384-well Plate
Quantity
2500 Assays
Type
Competitor Assay Kit
Contents & Storage
Contains:
• 4 tubes full-length ER Alpha Recombinant Protein (store at -68 to -85°C) • 1 tube of Fluormone ES2 Green Tracer (store at -5 to -30°C) • 2 bottles of ES2 Screening Buffer (store at -5 to -30°C)