Thermo Scientific Chemicals

3-Bromo-o-xylene, 99%, Thermo Scientific Chemicals

Catalog number: A15889.06
5 g, Each
Thermo Scientific Chemicals

3-Bromo-o-xylene, 99%, Thermo Scientific Chemicals

Catalog number: A15889.06
5 g, Each
Quantity
Catalog number: A15889.06
also known as A15889-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
576-23-8
IUPAC Name
1-bromo-2,3-dimethylbenzene
Molecular Formula
C8H9Br
InChI Key
WLPXNBYWDDYJTN-UHFFFAOYSA-N
SMILES
CC1=CC=CC(Br)=C1C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Refractive Index
1.5595-1.5625 @ 20?C
Form
Liquid

Description

3-Bromo-o-xylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-o-xylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text