Thermo Scientific Chemicals

3,4-Dihydroxyphenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A15893.06
5 g, Each
Thermo Scientific Chemicals

3,4-Dihydroxyphenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A15893.06
5 g, Each
Quantity
Catalog number: A15893.06
also known as A15893-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102-32-9
IUPAC Name
2-(3,4-dihydroxyphenyl)acetic acid
Molecular Formula
C8H8O4
InChI Key
CFFZDZCDUFSOFZ-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(O)C(O)=C1
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point
127-132?C
Appearance (Color)
White to cream to grey to pale brown to purple-grey
Clarity
0.5% w/v solution in ethanol will be clear to very faintly turbid by JIS K8001 5.2.
Form
Powder

Description

3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.

Solubility
Soluble in water and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Air sensitive. Incompatible with strong bases and strong oxidizing agents. Store at room temperature
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text