Thermo Scientific Chemicals

DL-Norvaline, 98%, Thermo Scientific Chemicals

Catalog number: A15900.36
500 g, Each
Thermo Scientific Chemicals

DL-Norvaline, 98%, Thermo Scientific Chemicals

Catalog number: A15900.36
500 g, Each
Quantity
Catalog number: A15900.36
also known as A15900-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
760-78-1
IUPAC Name
2-aminopentanoic acid
Molecular Formula
C5H11NO2
InChI Key
SNDPXSYFESPGGJ-UHFFFAOYSA-N
SMILES
CCCC(N)C(O)=O
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White

Description

DL-Norvaline is a neurotransmitter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Norvaline is a neurotransmitter.

Solubility
Soluble in water.

Notes
Store in cool dry place.. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text