Thermo Scientific Chemicals

4-Nitrophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15901.14
25 g, Each
Thermo Scientific Chemicals

4-Nitrophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15901.14
25 g, Each
Quantity
Catalog number: A15901.14
also known as A15901-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-03-0
IUPAC Name
2-(4-nitrophenyl)acetic acid
Molecular Formula
C8H7NO4
InChI Key
YBADLXQNJCMBKR-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(C=C1)[N+]([O-])=O
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
149-156?C
Appearance (Color)
White to cream to pale brown or yellow

Description

It is an important raw material and intermediate used in organic synthesis and also used for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis and also used for medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text