It is an important raw material and intermediate used in organic synthesis and also used for medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis and also used for medicine.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only
J. Herman. Schauble.; Elisabeth. Hertz. Reinvestigation of the Mannich reaction of 4-nitrophenylacetic acid and 2,4-dinitrophenylacetic acid. J. Org. Chem. 1970, 35 (8),2529-2532 .
Ioakim K. Spiliopoulos.; John A. Mikroyannidis. Rigid-Rod Polyamides and Polyimides Derived from 4,3‘‘-Diamino-2‘,6‘-diphenyl- or Di(4-biphenylyl)-p-terphenyl and 4-Amino-4‘‘-carboxy-2‘,6‘-diphenyl-p-terphenyl. Macromolecules. 1998, 31 (2),522-529 .