Thermo Scientific Chemicals

3-Acetamidobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15902.22
100 g, Each
Thermo Scientific Chemicals

3-Acetamidobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15902.22
100 g, Each
Quantity
Catalog number: A15902.22
also known as A15902-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
587-48-4
IUPAC Name
sodium 3-acetamidobenzoate
Molecular Formula
C9H8NNaO3
InChI Key
FEMJCNCAWASVDS-UHFFFAOYSA-M
SMILES
[Na+].CC(=O)NC1=CC=CC(=C1)C([O-])=O
Assay (Silylated GC)
≥97.5%
Appearance (Color)
Pale cream to cream to pale brown
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
248.0-255.0?C
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text