Thermo Scientific Chemicals

Isophthaloyl dichloride, 98%, Thermo Scientific Chemicals

Catalog number: A15904.36
500 g, Each
Thermo Scientific Chemicals

Isophthaloyl dichloride, 98%, Thermo Scientific Chemicals

Catalog number: A15904.36
500 g, Each
Quantity
Catalog number: A15904.36
also known as A15904-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-63-8
IUPAC Name
benzene-1,3-dicarbonyl dichloride
Molecular Formula
C8H4Cl2O2
InChI Key
FDQSRULYDNDXQB-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC(=CC=C1)C(Cl)=O
Appearance (Color)
White or colorless
Assay from Supplier's CofA
≥97.5 to ≤102.5%
Freezing Point
42-45°C
Form
Liquid, fused solid, or crystalline powder

Description

Isophthaloyl dichloride is used in a variety of performance polymers and fibers, where it impacts flame resistance, temperature stability, chemical resistance, and flexibility. In addition, it is an effective stabilizer for urethane prepolymers due to its ability to scavenge water. Also used in aromatic fiber raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophthaloyl dichloride is used in a variety of performance polymers and fibers, where it impacts flame resistance, temperature stability, chemical resistance, and flexibility. In addition, it is an effective stabilizer for urethane prepolymers due to its ability to scavenge water. Also used in aromatic fiber raw materials.

Solubility
Reacts with water.

Notes
Incompatible with strong bases and strong oxidizing agents. It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text