Thermo Scientific Chemicals

Aurintricarboxylic acid, Thermo Scientific Chemicals

Catalog number: A15905.14
25 g, Each
Thermo Scientific Chemicals

Aurintricarboxylic acid, Thermo Scientific Chemicals

Catalog number: A15905.14
25 g, Each
Quantity
Catalog number: A15905.14
also known as A15905-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4431-00-9
IUPAC Name
triammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Molecular Formula
C22H25N3O9
InChI Key
UDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES
[NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
Form
Powder or crystalline powder
Comment
Suitable for the estimation of aluminium
Identification (FTIR)
Conforms
Appearance (Color)
Dark red
Assay (Aqueous acid-base Titration)
≥90%

Description

Aurintricarboxylic acid is employed as a powerful inhibitor of cellular processes, including apoptosis, that are dependent on the formation of protein-nucleic acid complexes. It binds to a FGF and reduces its angiogenic activity. Aurintricarboxylic Acid has been shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. It Inhibits protein-nucleic acid interactions. It stimulates the tyrosine phosphorylation of MAP kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aurintricarboxylic acid is employed as a powerful inhibitor of cellular processes, including apoptosis, that are dependent on the formation of protein-nucleic acid complexes. It binds to a FGF and reduces its angiogenic activity. Aurintricarboxylic Acid has been shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. It Inhibits protein-nucleic acid interactions. It stimulates the tyrosine phosphorylation of MAP kinases.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text