Thermo Scientific Chemicals

Methyl benzilate, 98%, Thermo Scientific Chemicals

Catalog number: A15907.30
250 g, Each
Thermo Scientific Chemicals

Methyl benzilate, 98%, Thermo Scientific Chemicals

Catalog number: A15907.30
250 g, Each
Quantity
Catalog number: A15907.30
also known as A15907-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
76-89-1
IUPAC Name
methyl 2-hydroxy-2,2-diphenylacetate
Molecular Formula
C15H14O3
InChI Key
LJFIHTFNTGQZJL-UHFFFAOYSA-N
SMILES
COC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
71.0-77.0?C

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text