Thermo Scientific Chemicals

Nonanal, 97%, Thermo Scientific Chemicals

Catalog number: A15908.22
100 g, Each
Thermo Scientific Chemicals

Nonanal, 97%, Thermo Scientific Chemicals

Catalog number: A15908.22
100 g, Each
Quantity
Catalog number: A15908.22
also known as A15908-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-19-6
IUPAC Name
nonanal
Molecular Formula
C9H18O
InChI Key
GYHFUZHODSMOHU-UHFFFAOYSA-N
SMILES
CCCCCCCCC=O
Form
Liquid
Free acid (titration)
≤1.5%
Refractive Index
1.4215-1.4265 @ 20?C
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

Nonanal is used for fragranced end-products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonanal is used for fragranced end-products.

Solubility
Insoluble in water.

Notes
Incompatible with reducing agents, strong basesand strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text