Nonanal is used for fragranced end-products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nonanal is used for fragranced end-products.
Solubility
Insoluble in water.
Notes
Incompatible with reducing agents, strong basesand strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only
Arno Behr.; Dietmar Obst.; Barbara Turkowski. Isomerizing hydroformylation of trans-4-octene to n-nonanal in multiphase systems: acceleration effect of propylene carbonate. Journal of Molecular Catalysis A: Chemical 2005, 226 (2), 215-219.
Julia Hurst Bowman.; Dennis J. Barket, Jr.; Paul B. Shepson. Atmospheric Chemistry of Nonanal. Environ. Sci. Technol. 2003, 37 (10), 2218-2225.