Thermo Scientific Chemicals

4-Bromobenzenesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15909.22
100 g, Each
Thermo Scientific Chemicals

4-Bromobenzenesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15909.22
100 g, Each
Quantity
Catalog number: A15909.22
also known as A15909-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-58-8
IUPAC Name
4-bromobenzene-1-sulfonyl chloride
Molecular Formula
C6H4BrClO2S
InChI Key
KMMHZIBWCXYAAH-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=CC=C(Br)C=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
72.0-78.0?C
Appearance (Color)
White to cream to pale brown

Description

4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.

Solubility
Soluble in Tetrahydrofuran, petroleum ether and 1,4-dioxane. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text