Thermo Scientific Chemicals

3-(Trifluoromethyl)aniline, 99%, Thermo Scientific Chemicals

Catalog number: A15910.22
100 g, Each
Thermo Scientific Chemicals

3-(Trifluoromethyl)aniline, 99%, Thermo Scientific Chemicals

Catalog number: A15910.22
100 g, Each
Quantity
Catalog number: A15910.22
also known as A15910-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-16-8
IUPAC Name
3-(trifluoromethyl)aniline
Molecular Formula
C7H6F3N
InChI Key
VIUDTWATMPPKEL-UHFFFAOYSA-N
SMILES
NC1=CC=CC(=C1)C(F)(F)F
Appearance (Color)
Clear colorless to yellow to orange
Assay (GC)
≥98.5%
Refractive Index
1.4785-1.4815 @ 20°C
Form
Liquid
Identification (FTIR)
Conforms

Description

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water. [5 g/L (20°C)]

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text