Thiophenol, 99+%, Thermo Scientific Chemicals
Thiophenol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiophenol, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
50 g
250 g
Catalog number A15916.0B
also known as A15916-0B
Price (USD)/ Each
204.65
Online exclusive
227.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
204.65
Online exclusive
227.00 
Save 22.35 (10%)
Add to cart
Thiophenol, 99+%, Thermo Scientific Chemicals
Catalog numberA15916.0B
Price (USD)/ Each
204.65
Online exclusive
227.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS108-98-5
IUPAC Namebenzenethiol
Molecular FormulaC6H6S
InChI KeyRMVRSNDYEFQCLF-UHFFFAOYSA-N
SMILESSC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥99.0%
Refractive Index1.5880-1.5915 @ 20?C
Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

Solubility
Miscible with most organic solvents and aqueous base. Immiscible with water.

Notes
Incompatible with strong acids, bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. For a detailed investigation of alkylation using, e.g. K2CO3 in acetone, see: Synth. Commun., 22, 1691 (1992). Unactivated aryl chlorides undergo nucleophilic substitution in the presence of K2CO3 in NMP to give aryl sulfides: J. Org. Chem., 56, 862 (1991).
  2. Michael additions occur readily with electron deficient alkenes. Subsequent chlorination with NCS and dehydrochlorination provides a stereoselective route to vinyl thioethers: J. Org. Chem., 46, 235 (1981):
  3. The Cu(I) derivative, phenylthiocopper forms complexes with alkyllithiums, which are useful alkyl transfer reagents, undergoing conjugate addition to enones, coupling with alkyl iodides to alkanes, and converting acid halides to ketones: J. Am. Chem. Soc., 95, 7788 (1973); Synthesis, 662 (1974); Org. Synth. Coll., 6, 248 (1988).
  4. Chlorination of PhSH with NCS: J. Org. Chem., 37, 1367 (1972), or SO2Cl2: Org. Synth. Coll., 8, 550 (1993), gives the useful reagent benzenesulfenyl chloride, PhSCl.
  5. Dilithiation of PhSH with n-BuLi and TMEDA gives exclusive ortho-metallation: J. Am. Chem. Soc., 111, 654, 665, 2327 (1989). Subsequent treatment of the dilithio species with DMF, followed by in situ reaction with chloroacetone provides a useful route to the benzo[b]thiophene system: J. Org. Chem., 58, 1293 (1993):
  6. Interception of the DMF adduct provides a source of the unstable 2-mercaptobenzaldehyde. Alternatively, ortho-metallated thiophenols can be formed by lithiation of the THP ether, in which coordination by oxygen favors ortho-lithiation: J. Am. Chem. Soc., 111, 658 (1989).