Thermo Scientific Chemicals

Thiophenol, 99+%, Thermo Scientific Chemicals

Catalog number: A15916.30
250 g, Each
Thermo Scientific Chemicals

Thiophenol, 99+%, Thermo Scientific Chemicals

Catalog number: A15916.30
250 g, Each
Quantity
Catalog number: A15916.30
also known as A15916-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-98-5
IUPAC Name
benzenethiol
Molecular Formula
C6H6S
InChI Key
RMVRSNDYEFQCLF-UHFFFAOYSA-N
SMILES
SC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥99.0%
Refractive Index
1.5880-1.5915 @ 20?C

Description

Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

Solubility
Miscible with most organic solvents and aqueous base. Immiscible with water.

Notes
Incompatible with strong acids, bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text