1-Bromo-4-n-hexylbenzene, 97%, Thermo Scientific Chemicals
1-Bromo-4-n-hexylbenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-4-n-hexylbenzene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A15918.14
also known as A15918-14
Price (USD)/ Each
442.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
442.00
Add to cart
1-Bromo-4-n-hexylbenzene, 97%, Thermo Scientific Chemicals
Catalog numberA15918.14
Price (USD)/ Each
442.00
-
Add to cart
Chemical Identifiers
CAS23703-22-2
IUPAC Name1-bromo-4-hexylbenzene
Molecular FormulaC12H17Br
InChI KeyMXHOLIARBWJKCR-UHFFFAOYSA-N
SMILESCCCCCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
Refractive Index1.5185-1.5235 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only