Thermo Scientific Chemicals

2,3-Dimethoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Catalog number: A15936.30
250 g, Each
Thermo Scientific Chemicals

2,3-Dimethoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Catalog number: A15936.30
250 g, Each
Quantity
Catalog number: A15936.30
also known as A15936-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7345-82-6
IUPAC Name
3-(2,3-dimethoxyphenyl)prop-2-enoic acid
Molecular Formula
C11H12O4
InChI Key
QAXPUWGAGVERSJ-UHFFFAOYSA-N
SMILES
COC1=CC=CC(C=CC(O)=O)=C1OC
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.0%
Melting Point (clear melt)
177.0-184.0?C
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2E,2E)-N,N,N-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2′E,2′′E)-N,N′,N′′-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text