Triphenylamine is used in organic light-emitting diode as hole-transporters and as a pharmaceutical intermediate. It is coated on film bases as primary photoconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triphenylamine is used in organic light-emitting diode as hole-transporters and as a pharmaceutical intermediate. It is coated on film bases as primary photoconductor.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
A. Nozomu Tamoto.; Chihaya Adachi.; Kazukiyo Nagai. Electroluminescence of 1,3,4-Oxadiazole and Triphenylamine-Containing Molecules as an Emitter in Organic Multilayer Light Emitting Diodes. Chem. Mater. 1997, 9 (5), 077-1085.
M. Malagoli.; J.L. Brédas. Density functional theory study of the geometric structure and energetics of triphenylamine-based hole-transporting molecules. Chemical Physics Letters. 2000, 327 (1-2), 3-17.