Thermo Scientific Chemicals

Diacetone acrylamide, 99%, Thermo Scientific Chemicals

Catalog number: A15940.18
50 g, Each
Thermo Scientific Chemicals

Diacetone acrylamide, 99%, Thermo Scientific Chemicals

Catalog number: A15940.18
50 g, Each
Quantity
Catalog number: A15940.18
also known as A15940-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2873-97-4
IUPAC Name
N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide
Molecular Formula
C9H15NO2
InChI Key
OMNKZBIFPJNNIO-UHFFFAOYSA-N
SMILES
CC(=O)CC(C)(C)NC(=O)C=C
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Form
Fused solid or flakes
Appearance (Color)
White to cream

Description

Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.

Solubility
Soluble in water, methanol, benzene and tetrahydrofuran.

Notes
Store in cool place. Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text