Thermo Scientific Chemicals

4-Bromobenzotrifluoride, 99%, Thermo Scientific Chemicals

Catalog number: A15942.06
5 g, Each
Thermo Scientific Chemicals

4-Bromobenzotrifluoride, 99%, Thermo Scientific Chemicals

Catalog number: A15942.06
5 g, Each
Quantity
Catalog number: A15942.06
also known as A15942-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
402-43-7
IUPAC Name
1-bromo-4-(trifluoromethyl)benzene
Molecular Formula
C7H4BrF3
InChI Key
XLQSXGGDTHANLN-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(Br)C=C1
Form
Crystals or powder or crystalline powder or clear liquid as melt
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Refractive Index
1.4715-1.4745 @ 20?C
Appearance (Color)
Colorless to white to pale yellow

Description

4-Bromobenzotrifluoride is used to study the catalytic activity of palladacycle using Suzuki-Miyaura reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzotrifluoride is used to study the catalytic activity of palladacycle using Suzuki-Miyaura reaction.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which areopened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text