4-Bromobenzotrifluoride is used to study the catalytic activity of palladacycle using Suzuki-Miyaura reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromobenzotrifluoride is used to study the catalytic activity of palladacycle using Suzuki-Miyaura reaction.
Solubility
Not miscible or difficult to mix.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which areopened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only
Woen Susanto.; Chi-Yuan Chu.; Wei Jie Ang,.; Tzyy-Chao Chou.; Lee-Chiang Lo.; Yulin Lam. Development of a fluorous, oxime-based palladacycle for microwave-promoted carbon-carbon coupling reactions in aqueous media. Green Chem. 2012, 14 77-80 .
Mark Sundermeier Dr., Alexander Zapf Dr.; Matthias Beller Prof. Dr. A Convenient Procedure for the Palladium-Catalyzed Cyanation of Aryl Halides. Angewandte Chemie International Edition. 2003, 42 (14), 1661-1664.