Thermo Scientific Chemicals

3-Cyanobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A15943.06
5 g, Each
Thermo Scientific Chemicals

3-Cyanobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A15943.06
5 g, Each
Quantity
Catalog number: A15943.06
also known as A15943-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24964-64-5
IUPAC Name
3-formylbenzonitrile
Molecular Formula
C8H5NO
InChI Key
HGZJJKZPPMFIBU-UHFFFAOYSA-N
SMILES
O=CC1=CC=CC(=C1)C#N
Melting Point (clear melt)
73.0-82.0?C
Free acid (titration)
≤1.5%
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder or granules
Appearance (Color)
White to cream to yellow

Description

3-Cyanobenzaldehyde is used in the synthesis of 3-(6,6-dimethyl-5,6-dihydro-4H-benzo[7,8]chromeno[6,5-d]oxazol-2-yl)benzonitrile. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyanobenzaldehyde is used in the synthesis of 3-(6,6-dimethyl-5,6-dihydro-4H-benzo[7,8]chromeno[6,5-d]oxazol-2-yl)benzonitrile. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text