4-Methoxybenzonitrile is used as raw material of parfum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzonitrile is used as raw material of parfum.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
RUO – Research Use Only
General References:
- Boris Schmidt.; Daniela Meid.; Daniel Kieser. Safe and fast tetrazole formation in ionic liquids. Tetrahedron. 2007, 63 (2), 492-496.
- R.K. Goel.; M.L. Agarwal. Electronic and vibrational spectra and thermodynamic functions of 3- and 4-methoxy benzonitriles. Spectrochimica Acta Part A: Molecular Spectroscopy. 1982, 38 (6), 583-590.