Thermo Scientific Chemicals

1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15947.22
100 g, Each
Thermo Scientific Chemicals

1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15947.22
100 g, Each
Quantity
Catalog number: A15947.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
554-95-0
Assay (HPLC)
≥97.5%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

Solubility
Soluble in water, ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text