1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
100 g
250 g
Catalog number A15947.36
Price (USD)/ Each
256.65
Online exclusive
285.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
256.65
Online exclusive
285.00 
Save 28.35 (10%)
Add to cart
1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
Catalog numberA15947.36
Price (USD)/ Each
256.65
Online exclusive
285.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS554-95-0
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

Solubility
Soluble in water, ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Choi, Y.; Kim, Y. R.; Kang, Y. S.; Kang, S. W. Enhanced CO2 separation performance of polymer composite membranes through the synergistic effect of 1,3,5-benzenetricarboxylic acid. Chem. Eng. J. 2015, 279, 273-276.
  2. Liu, W.; He, H.; Song, Y.; Du, J.; Sun, F. Solvent-induced generation of two cadmium-based metal-organic frameworks from 1,3,5-benzenetricarboxylic acid ligand. Inorg. Chem. Commun. 2015, 57, 11-14.