Decafluorobiphenyl is used as an intermediate in organic synthesis and in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Decafluorobiphenyl is used as an intermediate in organic synthesis and in pharmaceuticals.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
Frank Mercer.; Tim Goodman.; Janusz Wojtowicz.; David Duff. Synthesis and characterization of fluorinated aryl ethers prepared from decafluorobiphenyl. Journal of Polymer Science Part A: Polymer Chemistry. 1992, 30 (8), 1767-1770 .
N. Boden.; J.W. Emsley.;J. Feeney.; L.H. Sutcliffe. The N.M.R. spectrum and configuration of decafluorobiphenyl. Molecular Physics: An International Journal at the Interface Between Chemistry and Physics. 1964, 8 (5), 467-471.