Thermo Scientific Chemicals

Decafluorobiphenyl, 99%, Thermo Scientific Chemicals

Catalog number: A15948.14
25 g, Each
Thermo Scientific Chemicals

Decafluorobiphenyl, 99%, Thermo Scientific Chemicals

Catalog number: A15948.14
25 g, Each
Quantity
Catalog number: A15948.14
also known as A15948-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
434-90-2
IUPAC Name
decafluoro-1,1'-biphenyl
Molecular Formula
C12F10
InChI Key
ONUFSRWQCKNVSL-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
65.0-72.0?C

Description

Decafluorobiphenyl is used as an intermediate in organic synthesis and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decafluorobiphenyl is used as an intermediate in organic synthesis and in pharmaceuticals.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text