Thermo Scientific Chemicals

Thiomorpholine, 98%, Thermo Scientific Chemicals

Catalog number: A15958.14
25 g, Each
Thermo Scientific Chemicals

Thiomorpholine, 98%, Thermo Scientific Chemicals

Catalog number: A15958.14
25 g, Each
Quantity
Catalog number: A15958.14
also known as A15958-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-90-0
IUPAC Name
hydrogen thiomorpholine chloride
Molecular Formula
C4H10ClNS
InChI Key
QSJAHJYXDRUZMY-UHFFFAOYSA-N
SMILES
[H+].[Cl-].C1CSCCN1
Assay (GC)
>97.5%
Form
Liquid
Refractive Index
1.5370-1.5410 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

Solubility
Miscible with waterMiscible with water and organic solvents.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text